Cole-Parmer Spex® 3644-450 UltraBind® Binder Powder, <20 µm, 450 g
Representative image only.
Cole-Parmer Item # EW-04577-51
Prepare samples for reproducible and contamination-free results
  • Select binders from powder, tablet, or liquid form depending on your application
  • UltraBind powder with a particle size of <20 µm
  • 450 g bottle
  • Will bond 600 to 750 sample disks or provide matrix for 75 to 105 supported sample disks
$312.00USD / Each
Don't forget to order scoops
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPowder
  • DescriptionUltraBind® Binder Powder, <20 µm, 450 g
Key Features
  • Select binders from powder, tablet, or liquid form depending on your application
  • UltraBind powder with a particle size of <20 µm
  • 450 g bottle
  • Will bond 600 to 750 sample disks or provide matrix for 75 to 105 supported sample disks
More About this Item
High-quality sample preparation is the key to achieving consistent and reliable analytical results. Powders are ideal for samples that crumble or ablate after pressing. They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks. Tablets are an efficient way to add binder to XRF sample disks. Liquid easily mixes with the sample.
Specially formulated blend combines the moisture resistance of paraffin with the ease of removing a prepared disk of the cellulose binder. Blends quickly and completely with samples at 10 to 25% by weight to form XRF sample disks or use undiluted as a sample matrix with sleeve-and-plunger set 04577-38 or 04577-45. UltraBind powder may include low levels of NaCl.