Spex SamplePrep 6754 Vial Extractor/End Plug Remover for Small Grinding Vials
Representative image only.
Spex SamplePrep Item # EW-04576-31
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Lifts small grinding vials in and out of the Freezer/Mill® cryogenic grinders
  • Lever assists with end plug extraction
  • 1 year warranty
PRODUCT OPTIONS
$554.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity1 small vial
  • DescriptionVial Extractor/End Plug Remover for Small Grinding Vials
  • Warranty1 year
1 Year Warranty
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Lifts small grinding vials in and out of the Freezer/Mill® cryogenic grinders
  • Lever assists with end plug extraction
  • 1 year warranty
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Makes it easier to open cold grinding vials immediately after they have been removed from the Freezer/Mill. Compatible with small grinding vial sets 04576-34, -35, and 61043-07.
Compatible with the vial holder used in the microvial set 61043-06.
Included with the purchase of each 6775 Freezer/Mill®
Included with the purchase of each 6870S Small Grinding Vial Accessory Package.