Spex SamplePrep 6761E Replacement End Plugs for Small Grinding Set 04576-34, PC; 2/PK
Representative image only.
Spex SamplePrep Item # EW-61034-58
Keep spare items to ensure there is no down time
  • Suitable for use with the Spex® SamplePrep Freezer/Mill® grinding vial sets
  • Polycarbonate end plugs
  • Eliminate metal contamination
  • Ideal for grinding soft plant and animal tissues
  • Pack of 2
PRODUCT OPTIONS
$249.00USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Compatible WithSmall grinding vial set 04576-34
  • DescriptionReplacement End Plugs for Small Grinding Set 04576-34, PC; 2/PK
  • Warranty1 year
1 Year Warranty
Key Features
  • Suitable for use with the Spex® SamplePrep Freezer/Mill® grinding vial sets
  • Polycarbonate end plugs
  • Eliminate metal contamination
  • Ideal for grinding soft plant and animal tissues
  • Pack of 2
  • 1 year warranty
More About this Item
Optimize performance of your Spex® SamplePrep Freezer/Mill® grinding vial sets with these replacement parts and accessories.