Spex SamplePrep 6781S Grinding Vial Set, Small Vial, 0.5 to 4 mL, SS Cylinder, End plugs, and Impactor

Representative image only.
Spex SamplePrep Item # EW-04577-80
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Vial sets consist of a center cylinder, two end plugs, and an impactor
  • Ideal for when contact with plastic is not recommended
  • Easy to clean and reusable

PRODUCT OPTIONS

$534.00USD / Pkg of 1

SPECS & DESCRIPTION

Specifications & Description

  • Use with GeneratorsFreezer/Mill® cryogenic grinders 04500-39 to -42 and 04577-77, -92 to -94
  • Min Sample Size (mL)0.5
  • Min Sample Size (g)0.5
  • Max Sample Size (mL)4
  • Max Sample Size (g)5
  • MaterialSS cylinder, end plugs with O-ring, and impactor
  • DescriptionGrinding Vial Set, Small Vial, 0.5 to 4 mL, SS Cylinder, End plugs, and Impactor
  • Warranty1 year
1 Year Warranty

Includes

Ones et of stainless steel (SS) end plugs, one SS impactor, one SS cylinder, and 10 O-rings.

Key Features

  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Vial sets consist of a center cylinder, two end plugs, and an impactor
  • Ideal for when contact with plastic is not recommended
  • Easy to clean and reusable
  • Sample capacity of 0.1 to 5 g
  • 1 year warranty

More About this Item

Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.

The closed grinding vial prevents sample cross-contamination, retains volatiles, and protects hazardous and critical samples. All stainless steel material makes it ideal for grinding hard samples like bone and circuit boards. End plugs, impactor, and cylinder are autoclavable and reusable. Grinding vial set is compatible with 6775 Freezer/Mill® cryogenic grinder. For use with 6875, 6875A, and 6875D Freezer/Mill cryogenic grinders, order multivial holder 04500-51. Easily remove end plugs using extractor 04550-46.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat