Cole-Parmer Spex® 6791 Vial Impactor Extractor
Representative image only.
Cole-Parmer Item # EW-04500-53
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Easily retrieve impactors from vials
PRODUCT OPTIONS
$20.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)50
  • DescriptionVial Impactor Extractor
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Easily retrieve impactors from vials
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Vial impactor for Freezer/Mill® Vials.