Spex SamplePrep 6804 Vial Extractor/End Plug Remover for Mid-Size and Large Grinding Vials
Representative image only.
Spex SamplePrep Item # EW-04576-38
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Lifts large and mid-size grinding vials in and out of the Freezer/Mill® cryogenic grinders
  • Lever assists with end plug extraction
  • 1 year warranty
PRODUCT OPTIONS
$668.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity1 large vial
  • DescriptionVial Extractor/End Plug Remover for Mid-Size and Large Grinding Vials
  • Warranty1 year
1 Year Warranty
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Lifts large and mid-size grinding vials in and out of the Freezer/Mill® cryogenic grinders
  • Lever assists with end plug extraction
  • 1 year warranty
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Makes it easier to open cold grinding vials immediately after they have been removed from the Freezer/Mill® cryogenic grinders. Compatible with large grinding vial sets 61043-09 and -11. Compatible with mid-size vial sets 61043-08 and -12 when used with vial adapter 04576-42.
Compatible with the following mid-size vials when used in conjunction with the 6884 Mid-Size Grinding Vial Adapter: 6881 Mid-Size Grinding Vial Set, 6883 Mid-Size Cr-Free Grinding Vial Set, 6885 Mid-Size Poly-Vial Set
Included with the purchase of each 6870L Large Grinding Vial Accessory Package.