Spex SamplePrep 6805 Rack for Large Grinding Vials, 6 Vial Capacity
Representative image only.
Spex SamplePrep Item # EW-04576-39
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Durable epoxy-coated steel construction
  • 1 year warranty
PRODUCT OPTIONS
$100.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity6 large vials
  • DescriptionRack for Large Grinding Vials, 6 Vial Capacity
  • Warranty1 year
1 Year Warranty
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Durable epoxy-coated steel construction
  • 1 year warranty
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Holds up to six large grinding vials 61043-09 and -11.