Spex SamplePrep 6808M Mid-Size Vial Adapter Kit for End Plug Extractor with Lever 04500-50
Representative image only.
Spex SamplePrep Item # EW-04500-48
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with extractor with lever 04500-50
PRODUCT OPTIONS
$142.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)25
  • Tubes Held1
  • DescriptionMid-Size Vial Adapter Kit for End Plug Extractor with Lever 04500-50
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with extractor with lever 04500-50
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
When attached to the extractor 04500-50, it removes the end plugs from mid-size vials.