Spex SamplePrep 6870L Grinding Vial Accessory Package, Large
Representative image only.
Spex SamplePrep Item # EW-04577-87
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with grinding vial sets 61043-09 and -11
  • 1 year warranty
PRODUCT OPTIONS
$756.00USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGrinding Vial Accessory Package, Large
  • Warranty1 year
1 Year Warranty
Includes
One vial extractor/opener 04576-38 and vial rack 04576-39.
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with grinding vial sets 61043-09 and -11
  • 1 year warranty
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
This accessory package contains everything necessary to run large grinding vials 61043-09 and -11 in the 6875, 6875A, and 6875D Freezer/Mill® cryogenic grinders.