Spex SamplePrep 6870M-1 Grinding Vial Accessory Package, Mid-Size
Representative image only.
Spex SamplePrep Item # EW-04577-88
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with grinding vial sets 61043-08 and -12
  • 1 year warranty
PRODUCT OPTIONS
$634.00USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGrinding Vial Accessory Package, Mid-Size
  • Warranty1 year
1 Year Warranty
Includes
One one vial adapter 04576-44 and one vial adapter 04576-42 for extractor/end plug remover 04576-38.
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with grinding vial sets 61043-08 and -12
  • 1 year warranty
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
This accessory package contains everything needed to run mid-size grinding vials 61043-08 and -12 in 6875, 6875A, and 6875D Freezer/Mill® cryogenic grinders. NOTE: Mid-size vial adapter 04500- 52 (sold separately) must be used for mid-size poly-vial 61043-13 in 6875D Freezer/Mill® cryogenic grinder.