Spex SamplePrep 6870S Grinding Vial Accessory Package, Small
Representative image only.
Spex SamplePrep Item # EW-04577-89
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with grinding vial sets 04576-35 and 61043-07
  • 1 year warranty
PRODUCT OPTIONS
$834.00USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGrinding Vial Accessory Package, Small
  • Warranty1 year
1 Year Warranty
Includes
One multivial holder 04500-51, one vial extractor/opener 04576-31, and one vial rack 04576-32.
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with grinding vial sets 04576-35 and 61043-07
  • 1 year warranty
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
This accessory package is ideal for running small grinding vials in the 6875, 6875A, and 6875D Freezer/Mill® cryogenic grinders.