Spex SamplePrep 6884 Mid-Size Vial Adapter for Extractor 04576-38
Representative image only.
Spex SamplePrep Item # EW-04576-42
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with extractor 04576-38
  • 1 year warranty
PRODUCT OPTIONS
$188.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity1 mid-size vial
  • DescriptionMid-Size Vial Adapter for Extractor 04576-38
  • Warranty1 year
1 Year Warranty
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Compatible with extractor 04576-38
  • 1 year warranty
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
When attached to extractor 04576-38, it extracts mid-size grinding vials from cryogenic grinders. Use it to remove end plugs from mid-size grinding vial sets. Adapter is easily installed and removed.