Spex SamplePrep 6887 Vial Holder Adapter for Mid-Size Vial, Open-End
Representative image only.
Spex SamplePrep Item # EW-04576-44
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Adapter for mid-size vial sets 61043-08 and -12
  • 1 year warranty
PRODUCT OPTIONS
$444.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity1 mid-size vial
  • DescriptionVial Holder Adapter for Mid-Size Vial, Open-End
  • Warranty1 year
1 Year Warranty
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Adapter for mid-size vial sets 61043-08 and -12
  • 1 year warranty
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Use this holder with mid-size grinding vial sets 61043-08 and -12. It is open at the back end to allow the force of the impact to strike against the back anvil of the cryogenic grinder coil. Compatibel with 6875, 6875A, and 6875D Freezer/Mill® cryogenic grinders.