Cole-Parmer Spex® 6905 Cryogenic Dewar, Portable, 25 L; 1/EA
Representative image only.
Cole-Parmer Item # EW-04576-45
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Dispense liquid nitrogen directly into the Freezer/Mill® cryogenic grinders
  • Requires cryogenic transfer hose 04500-54 and 04576-46
  • Use for manual fill only
PRODUCT OPTIONS
$2,615.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity25 Liters
  • DescriptionCryogenic Dewar, Portable, 25 L; 1/EA
Includes
A liquid nitrogen withdrawal device.
Key Features
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Dispense liquid nitrogen directly into the Freezer/Mill® cryogenic grinders
  • Requires cryogenic transfer hose 04500-54 and 04576-46
  • Use for manual fill only
More About this Item
Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.
Store and dispense small amounts of liquid nitrogen directly into the Freezer/Mill® cryogenic grinders. Dewar features a sturdy, aluminum, five-wheel roller base making it safe and easy to move the it from room to room. Use the included withdrawal device to dispenses liquid nitrogen at rates up to 8 L/min.