Spex SamplePrep 8010 Multiple Sample Vial Adapter, Holds Seven 1/2" (12.7 mm) Dia Vials; 1/EA
Representative image only.
Spex SamplePrep Item # EW-04576-65
Use adapters to run multiple samples or to hold vial securely in place during grinding
  • Suitable for use with the Spex® SamplePrep high-energy Freezer/Mill® grinders
  • Simultaneously run seven vials in the High-Energy Mixer/Mill® Grinders
  • Excellent for processing multiple smaller samples
  • Compatible with vials 04575-87, 04575-89, 04575-90, 04577-20, and 04576-91
  • 1 year warranty
PRODUCT OPTIONS
$145.60USD / Each
$182.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum
  • Compatible With1/2" dia grinding vials
  • DescriptionMultiple Sample Vial Adapter, Holds Seven 1/2" (12.7 mm) Dia Vials; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Suitable for use with the Spex® SamplePrep high-energy Freezer/Mill® grinders
  • Simultaneously run seven vials in the High-Energy Mixer/Mill® Grinders
  • Excellent for processing multiple smaller samples
  • Compatible with vials 04575-87, 04575-89, 04575-90, 04577-20, and 04576-91
  • 1 year warranty
More About this Item
Optimize performance of your Spex® SamplePrep high-energy Freezer/Mill® grinder with a wide range of adapters and clamps.
Vials run at one time should be same length. Clamp adapter 04576-67 must be used when running vial 04575-87, 04575-89, 04577-20, or 04575-91.