Spex SamplePrep 8015 Clamps for Slurry Grinding in the Alumina Ceramic Grinding Vial Set 41005-86; 2/ST
Representative image only.
Spex SamplePrep Item # EW-04576-69
Use adapters to run multiple samples or to hold vial securely in place during grinding
  • Suitable for use with the Spex® SamplePrep high-energy Freezer/Mill® grinders
  • Clamps end caps to prevent leakage during sample grinding
  • One pair required for each vial set
  • 1 year warranty
PRODUCT OPTIONS
$150.40USD / Set of 1
$188.00USD / Set of 1
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithAlumina ceramic grinding vial set 41005-86
  • DescriptionClamps for Slurry Grinding in the Alumina Ceramic Grinding Vial Set 41005-86; 2/ST
  • Warranty1 year
1 Year Warranty
Key Features
  • Suitable for use with the Spex® SamplePrep high-energy Freezer/Mill® grinders
  • Clamps end caps to prevent leakage during sample grinding
  • One pair required for each vial set
  • 1 year warranty
More About this Item
Optimize performance of your Spex® SamplePrep high-energy Freezer/Mill® grinder with a wide range of adapters and clamps.