Cole-Parmer Spex® 8211S O-rings for 250 mL Grinding Jar; 10/Pk
Representative image only.
Cole-Parmer Item # EW-04900-04
Accessories to rapidly reduce hard and brittle samples with the BM-600 Planetary Mill
  • Jar rotation in combination with grinding balls impacting the sample and the walls of the jar allow for both milling and mixing
  • Excellent for grinding rocks, minerals, sand, cement, slag, ceramics, glass, and pharmaceuticals
PRODUCT OPTIONS
$52.50USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With250 mL Grinding Jar
  • DescriptionO-rings for 250 mL Grinding Jar; 10/Pk
Key Features
  • Jar rotation in combination with grinding balls impacting the sample and the walls of the jar allow for both milling and mixing
  • Excellent for grinding rocks, minerals, sand, cement, slag, ceramics, glass, and pharmaceuticals
More About this Item
These accessories are compatible with the Cole-Parmer® BM-600 Planetary Mills 04900-16 and 04900-17. Mill sold separately.
Choose from 250 mL or 500 mL jar capacities and stainless steel or hardened steel jars and grinding media. Generally, the grinding jar and grinding balls are made from the same material.