Cole-Parmer Spex® 8511-21 Handle Assembly for Large Hardened Steel Grinding Container
Representative image only.
Cole-Parmer Item # EW-41010-93
Easily convert your existing dish without handles
  • Optimize performance of your SM-300 ShatterBox®
  • Compatible with large hardened steel Grinding Container (41010-93)
PRODUCT OPTIONS
$393.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithLarge hardened steel container (04576-81)
  • DescriptionHandle Assembly for Large Hardened Steel Grinding Container
Made in the U.S.A.
Key Features
  • Optimize performance of your SM-300 ShatterBox®
  • Compatible with large hardened steel Grinding Container (41010-93)
More About this Item
The SM-300 ShatterBox® requires the use of a dish with a handle assembly. For existing dishes without handles, the corresponding handle assembly may be purchased and secured to the dish.