Cole-Parmer Spex® SM-300-230 ShatterBox® Verastile Ring and Puck Mill; 230 VAC, 50/60 Hz

Representative image only.
Cole-Parmer Item # EW-41010-84
Grind your samples with minimal loss in a soundproof enclosure
  • Store up to 10 different grinding programs
  • Robust IP65 rated industrial interface
  • Accommodates various grinding dishes
  • Sound-insulated steel cabinet
  • Electronically controlled, fully reproducible grinding methods with Ethernet capabilities
  • 3 year warranty

PRODUCT OPTIONS

$17,500.00USD / Each
Don't forget to order scoops

SPECS & DESCRIPTION

Specifications & Description

  • Speed (rpm)800 to 1200 (1050 for tungsten carbide dish)
  • Motor Size (hp)1/2
  • Power (VAC)230
  • Power (Hz)50/60
  • Height (in)54.3
  • Width (in)26.6
  • Height (cm)138
  • Width (cm)67.5
  • DescriptionShatterBox® Verastile Ring and Puck Mill; 230 VAC, 50/60 Hz
  • Warranty3 Year
3 Year WarrantyConformite EuropeenneMade in the U.S.A.

Includes

ShatterBox, instruction manual, USB memory stick, 10 mm wrench, Torx keys, and power cable.

Key Features

  • Store up to 10 different grinding programs
  • Robust IP65 rated industrial interface
  • Accommodates various grinding dishes
  • Sound-insulated steel cabinet
  • Electronically controlled, fully reproducible grinding methods with Ethernet capabilities
  • 3 year warranty

More About this Item

The Cole-Parmer® SM-300 ShatterBox® is a versatile ring and puck mill that can crush and grind many types and sizes of samples with minimal sample loss and increased safety. It features an automated locking dish and guided dish positioning with an automatic elevation of the dish for ease of use and greater flexibility. The versatility of sample containers allows for grinding within smaller dishes without the ring as well as the ability to increase sample sizes and capacity with the ring and puck system.

The ShatterBox swings a dish-shaped grinding container with a puck and ring inside in a tight high-speed circle. The sample is quickly crushed between the wall and floor of the container and the moving puck/ring grinding elements. Grinding times range from two to five minutes, with resultant particle size well below 100 µg/mL, and in some cases 5 µg/mL.

Typical samples include cement, superalloy, granite, glass, sand, slag, ceramics, seeds, grains, pigments, and catalyst supports.

The ShatterBox requires use of a dish with a handle assembly, which is available in hardened steel, tungsten carbide, alumina ceramic, and zirconia. Purchase grinding dish separately.

Effective September 2023, the Spex® SamplePrep 8550 ShatterBox® has now been rebranded Cole-Parmer® under model number SM-300 ShatterBox®. All the same great features and specifications, just with a new and improved look.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat

Bioz Article and Whitepaper Citations

Powered by Bioz See more details on Bioz