Spex SamplePrep Geno/Grinder® Nesting Tray Set for Stacking Microplates; 1/EA
Representative image only.
Spex SamplePrep Item # EW-04500-35
Get the most out of your Spex® SamplePrep homogenizer
  • Trays hold microplates in place
  • Set of two nesting mid-plates and one bottom plate
PRODUCT OPTIONS
$61.95USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Use with GeneratorsMicroplates and adjustable clamp 04605-08
  • DescriptionNesting Tray Set for Microplate Stacking; 1/EA
Includes
Two nesting mid-plates and one bottom plate.
Key Features
  • Trays hold microplates in place
  • Set of two nesting mid-plates and one bottom plate
More About this Item
Optimize your homogenizer with these quality accessories.
Nesting trays are used to stack microplates vertically in the adjustable clamp. The plates keep the samples secure during the grinding process.