Cole-Parmer Spex® 2151 Grinding Balls, Stainless Steel, 1/8" (3 mm); 1000/PK
Representative image only.
Cole-Parmer Item # EW-04500-20
Use grinding balls to grind samples in Geno/Grinder® homogenizers
  • Suitable for use with the Cole-Parmer® Geno/Grinder® homogenizers
  • Suitable for grinding small tissue samples
  • Autoclavable and reusable
  • Pack of 1,000 balls
PRODUCT OPTIONS
$206.75USD / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Material440C stainless steel
  • Compatible With96-well microplates 04575-64, 04576-90 and 04500-31, -32; PP microfuge tube 04606-90 or reinforced PP tube 04575-80
  • Diameter (in)0.125
  • Diameter (mm)3
  • DescriptionGrinding Balls, Stainless Steel, 1/8\" (3 mm); 1000/PK
Key Features
  • Suitable for use with the Cole-Parmer® Geno/Grinder® homogenizers
  • Suitable for grinding small tissue samples
  • Autoclavable and reusable
  • Pack of 1,000 balls
More About this Item
Optimize performance of your Geno/Grinder® homogenizers with a wide range of grinding ball options suitable for your application.
Constructed of 440C stainless steel, the 1/8" (3 mm) grinding balls are ideal for use with the Geno/Grinder® homogenizers.