Cole-Parmer Spex® 2150 Grinding Balls, Stainless Steel, 5/32" (4 mm); 5000/PK
Representative image only.
Cole-Parmer Item # EW-04500-19
Use grinding balls to grind samples in Geno/Grinder® homogenizers
  • Suitable for use with the Cole-Parmer® Geno/Grinder® homogenizers
  • Suitable for grinding small tissue samples
  • Compatible 96-well microplates
  • Autoclavable and reusable
  • Pack of 5,000 balls
PRODUCT OPTIONS
$921.00USD / Pkg of 5000
SPECS & DESCRIPTION
Specifications & Description
  • Material440C stainless steel
  • Compatible With96-well microplates 04500-31, 04500-32, 04575-64, and 04576-90
  • Diameter (in)0.15625
  • Diameter (mm)4
  • DescriptionGrinding Balls, Stainless Steel, 5/32\" (4 mm); 5000/PK
Key Features
  • Suitable for use with the Cole-Parmer® Geno/Grinder® homogenizers
  • Suitable for grinding small tissue samples
  • Compatible 96-well microplates
  • Autoclavable and reusable
  • Pack of 5,000 balls
More About this Item
Optimize performance of your Geno/Grinder® homogenizers with a wide range of grinding ball options suitable for your application.
Constructed of 440C stainless steel, the 5/32” (4 mm) grinding balls are compatible with 96-well microplates 04500-31, 04500-32, 04575-64, and 04576-90. Use with grinding ball dispenser 04500-18 to load titer plates with grinding balls in less than a second.