Spex SamplePrep Grinding Beads, Zirconia, 1/4" (6 mm); 1000/PK

Representative image only.
Spex SamplePrep Item # EW-04575-60
Use grinding balls to grind samples in Geno/Grinder® homogenizers
  • Ideal for when sample or solvent is not compatible with stainless steel

PRODUCT OPTIONS

$405.00USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • MaterialZirconia
  • Compatible WithReinforced 2 mL tube 04575-80
  • Diameter (in)0.25
  • Diameter (mm)6
  • DescriptionGrinding Beads, Zirconia, 1/4" (6 mm); 1000/PK

Key Features

  • Ideal for when sample or solvent is not compatible with stainless steel

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat