Cole-Parmer Spex® 2196-16-PE Homogenizer Foam Holder, 16 x 50 mL Tubes; 1/EA
Representative image only.
Cole-Parmer Item # EW-04500-27
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Ideal for grinding large samples
  • Process 16 samples at one time
PRODUCT OPTIONS
$64.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Tube Size (mL)50
  • Tubes Held16
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 16 x 50 mL Tubes; 1/EA
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Ideal for grinding large samples
  • Process 16 samples at one time
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.
Thick, rugged polyethylene foam block holds 16 standard (50 mL) conical bottom centrifuge tubes that are 115 mm in length.