Cole-Parmer Spex® 2257 Homogenizer Foam Holder, 16 x Grinding Vials 04575-73; 1/EA
Representative image only.
Cole-Parmer Item # EW-04575-75
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Polyethylene foam block holds sixteen vials 04575-73
PRODUCT OPTIONS
$200.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held16
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 16 x Grinding Vials 04575-73; 1/EA
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Polyethylene foam block holds sixteen vials 04575-73
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.
Thick, rugged foam block holds 24 standard 15 mL centrifuge tubes that are 119 mm in length.