Cole-Parmer Spex® 2259 Homogenizer Foam Holder, 2 x 750 mL Jars 04575-76; 1/EA
Representative image only.
Cole-Parmer Item # EW-04575-77
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Holds two jars 04575-76
PRODUCT OPTIONS
$160.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Tube Size (mL)750
  • Tubes Held2
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 2 x 750 mL Jars 04575-76; 1/EA
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Holds two jars 04575-76
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.