Cole-Parmer Spex® 1667 MiniG® Homogenizer Cryo-Block, Aluminum, 6 x 50 mL Round-Bottom Centrifuge Tubes; 1/EA
Representative image only.
Cole-Parmer Item # EW-04500-13
Accommodates a full range of sample vials with an adjustable clamp
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Aluminum vial holder helps maintain cold temperature during processing
  • Holds six 50 mL tubes
  • Hollow base makes it lightweight
PRODUCT OPTIONS
$449.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Use with GeneratorsMiniG® Homogenizer
  • MaterialAluminum
  • DescriptionMiniG® Homogenizer Cryo-Block, Aluminum, 6 x 50 mL Round-Bottom Centrifuge Tubes; 1/EA
Key Features
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Aluminum vial holder helps maintain cold temperature during processing
  • Holds six 50 mL tubes
  • Hollow base makes it lightweight
More About this Item
Multiple inserts are available for a variety of homogenizing applications.