Cole-Parmer Spex® 8018 Multi Vial Adapter for 2 mL Vial 04575-80, Aluminum
Representative image only.
Cole-Parmer Item # EW-04576-72
Use adapters to run multiple samples or to hold vial securely in place during grinding
  • Suitable for use with the Cole-Parmer® Freezer/Mill® Cryogenic Grinders
  • Simultaneously run seven 04575-80 vials in the High-Energy Mixer/Mill® Grinders
  • Excellent for processing multiple samples for biotechnology applications
  • Compatible with 2 mL plastic vials
PRODUCT OPTIONS
$190.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum
  • Compatible WithVial 04575-80
  • DescriptionMulti Vial Adapter for 2 mL Vial 04575-80, Aluminum
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® Cryogenic Grinders
  • Simultaneously run seven 04575-80 vials in the High-Energy Mixer/Mill® Grinders
  • Excellent for processing multiple samples for biotechnology applications
  • Compatible with 2 mL plastic vials
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinder with a wide range of adapters and clamps.