Cole-Parmer Spex® 8011 Multiple Sample Vial Adapter, Holds Four 3/4" (19.1 mm) Dia Vials; 1/EA
Representative image only.
Cole-Parmer Item # EW-04576-66
Use adapters to run multiple samples or to hold vial securely in place during grinding
  • Suitable for use with the Cole-Parmer® Freezer/Mill® Cryogenic Grinders
  • Simultaneously run four vials in the High-Energy Mixer/Mill® Grinders
  • Excellent for processing multiple smaller samples
  • Compatible with vials 04575-94, 04576-24, 04576-28, and 04577-64
PRODUCT OPTIONS
$191.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum
  • Compatible With3/4\" dia grinding vials
  • DescriptionMultiple Sample Vial Adapter, Holds Four 3/4\" (19.1 mm) Dia Vials; 1/EA
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® Cryogenic Grinders
  • Simultaneously run four vials in the High-Energy Mixer/Mill® Grinders
  • Excellent for processing multiple smaller samples
  • Compatible with vials 04575-94, 04576-24, 04576-28, and 04577-64
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinder with a wide range of adapters and clamps.
Vials run at one time should be same length. Clamp adapter 04576-67 must be used when running vial 04575-94.