Cole-Parmer Spex® 2254 Preloaded Vial Set, Polycarbonate, 75 mL, 2/PK

Representative image only.
Cole-Parmer Item # EW-04575-72
Preloaded with steel grinding balls for fast and convenient sample preparation
  • Optimize performance of your HG-400 MiniG® and Geno/Grinder® homogenizers
  • Each vial is loaded with two 1/2" (13 mm) steel grinding balls 04575-45
  • Ideal for soft to medium hard materials
  • Recommended mixing/grinding load is 10 to 30 g
  • Suitable for slurry grinding up to 40 mL

PRODUCT OPTIONS

$102.25USD / Pkg of 2

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)75
  • Tube MaterialPolycarbonate
  • Special FeaturesIncludes grinding balls
  • DescriptionPreloaded Vial Set, Polycarbonate, 75 mL, 2/PK

Includes

2 vials with caps and gasket and 2 steel balls.

Key Features

  • Optimize performance of your HG-400 MiniG® and Geno/Grinder® homogenizers
  • Each vial is loaded with two 1/2" (13 mm) steel grinding balls 04575-45
  • Ideal for soft to medium hard materials
  • Recommended mixing/grinding load is 10 to 30 g
  • Suitable for slurry grinding up to 40 mL

More About this Item

The 4, 5, and 15 mL vial sets come in a foam vial holder within a sturdy polyethylene carrier that has the same footprint and height as a deep-well titer plate. Carrier are used in pairs and fits adjustable clamp assembly 04605-08. Run up to four vial sets at a time for high-throughput tissue homogenization and cell lysis. Use frosted PE vials 04576-97 with Cryo-Block 04500-25 to preserve temperature sensitive samples during grinding. Polyethylene is compatible with commonly used reagents such as TRIzol® solution.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat