Cole-Parmer Spex® 6759 Replacement Holder for Microvial Grinding Set 61043-06; 1/EA
Representative image only.
Cole-Parmer Item # EW-04576-33
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Holds three microvial cylinders
PRODUCT OPTIONS
$303.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum
  • Compatible WithMicrovial grinding set 61043-06
  • DescriptionReplacement Holder for Microvial Grinding Set 61043-06; 1/EA
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Holds three microvial cylinders
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.