Cole-Parmer Spex® 6881P Replacement Impactor for Mid-Size Grinding Vial Set 61043-08, SS; 1/EA
Representative image only.
Cole-Parmer Item # EW-04577-98
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Stainless steel impactor
  • Autoclavable and reusable
PRODUCT OPTIONS
$146.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Compatible WithMid-size grinding vial set 61043-08
  • DescriptionReplacement Impactor for Mid-Size Grinding Vial Set 61043-08, SS; 1/EA
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Stainless steel impactor
  • Autoclavable and reusable
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.