Cole-Parmer Spex® 6753C Replacement Sample Extraction Tool for Microvial Grinding Set 61043-06; 1/EA
Representative image only.
Cole-Parmer Item # EW-04577-69
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Easily extract samples from microvials
PRODUCT OPTIONS
$64.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel spoke and silicone rubber plunger
  • Compatible WithMicrovial grinding set 61043-06
  • DescriptionReplacement Sample Extraction Tool for Microvial Grinding Set 61043-06; 1/EA
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Easily extract samples from microvials
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.
To use, simply inserted the extraction tool into an individual vial, then push it through to clear sample from the tube.