Cole-Parmer VapLock™ Air Inlet Valve, 1/4-28 UNF(M), 0.1-0.25 psi crack pressure, PTFE & FFKM; 1ea
Representative image only.
Cole-Parmer Item # EW-12020-61
Maintain pressure equilibrium within the bottle for safe solvent handling
  • Valves feature 0.1 PSI breaking pressure
PRODUCT OPTIONS
$70.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection Type1/4\"-28 UNF(M)
  • Breaking Pressure (PSI)0.1
  • Seal MaterialFFKM
  • Housing MaterialPTFE
  • Wetted MaterialsPTFE/FFKM
  • DescriptionInlet Check Valve, 1/4-28 UNF(M), 0.1-0.25 psi crack pressure, PTFE & FFKM; 1ea
Key Features
  • Valves feature 0.1 PSI breaking pressure
More About this Item
Valves minimize evaporation and relieve pressure to ensure safe delivery and containment of solvents. Air inlet check valves allow air to enter a bottle to displace vacuum while solvent is pumped out to an instrument.