Spex VapLock™ Complete Solvent Collection System, DOT waste container, PP manifold, 20 L
Representative image only.
Spex Item # EW-12020-67
Complete solution to safely collect solvent waste from laboratory instruments
  • Collect solvent waste from up to 9 effluent streams
  • Minimize exposure to harmful chemical vapors
  • DOT compliance for waste container: UN rating 1H1/Y1.8/100 for hazardous product and are FDA and USDA compliant
PRODUCT OPTIONS
$536.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)20
  • MaterialPolypropylene
  • Cap / Stopper Size70 mm Rieke 6TPI
  • Bottle ColorTransluscent
  • DescriptionComplete Solvent Collection System, 70mm Rieke 6TPI, PP manifold, 20 L
Includes
20-L translucent PE container (DOT approved), PP manifold with 70 mm Rieke 6TPI size cap, plugs for any unused ports, and an exhaust filter with color indicator.
Key Features
  • Collect solvent waste from up to 9 effluent streams
  • Minimize exposure to harmful chemical vapors
  • DOT compliance for waste container: UN rating 1H1/Y1.8/100 for hazardous product and are FDA and USDA compliant
More About this Item
Solvent waste systems come complete with everything you need to safely collect solvent waste from any system that generates solvent waste. Closed-system design reduces harmful and flammable solvent vapor emissions into the environment as well as leaks and spills.
Systems come with a 20-L polyethylene container with 70 mm Rieke 6TPI connection and a manifold with tubing connections for up to 9 waste lines. An easy-carry handle makes it easy to transport.