Cole-Parmer VapLock™ Conversion Kit for Normal Phase Solvents, Male and Female Quick Disconnects Compatible with 1/4" NPT(M) Manifold, 316 SS and PTFE; 1/EA
Representative image only.
Cole-Parmer Item # EW-12020-90
Use with Justrite® waste containers for handling aggressive solvents
  • 316 SS and PTFE components withstand aggresive solvents
$303.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Material316 SS fittings and PTFE O-ring
  • Compatible WithJustrite quick-disconnect disposal cans 86935-00 and 86395-10
  • DescriptionConversion Kit for Normal Phase Solvents, Male and Female Quick Disconnects Compatible with 1/4\" NPT(M) Manifold, 316 SS and PTFE; 1/EA
Key Features
  • 316 SS and PTFE components withstand aggresive solvents
More About this Item
Use the conversion kit exclusively for Justrite quick-disconnect disposal cans 86395-00 and -10. Plastic components that are included with models 86395-00 and -10 are incompatible with many aggresive solvents used in HPLC and will swell and leak. The conversion kit contains two 316 SS quick-disconnect fittings with PTFE O-rings to provide excellent chemical compatibility. Fittings feature 1/4" NPT(M) thread to screw into the Justrite container. Easily attach to Justrite container using two adjustable wrenches.