Cole-Parmer VapLock™ Fitting, Polypropylene, Straight, Compression to Threaded Adapter, 1/2" OD Tubing x 1/4" NPT(M); 1/EA
Representative image only.
Cole-Parmer Item # EW-12026-53
Designed for secure leak-resistant connections
  • Constructed of durable polypropylene
PRODUCT OPTIONS
$6.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeCompression x NPT(M)
  • Thread size NPT(M) (port 2) (in)1/4
  • Compression Size / Tube Od (Port 1) (in)1/2
  • Compression Size / Tube Od (Port 1) (mm)12.7
  • DescriptionFitting, Polypropylene, Straight, Compression to Threaded Adapter, 1/2\" OD Tubing x 1/4\" NPT(M); 1/EA
Key Features
  • Constructed of durable polypropylene
More About this Item
Adapters with large bore for semi-rigid tubing are constructed of two-piece molded polypropylene components and do not include a separate ferrule. Adapters withstand pressures up to 100 psi. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.