Spex VapLock™ Isolation Valve, 2-Way, NC, Side Mounted Ports
Representative image only.
Spex Item # EW-12021-20
Ideal for use with highly aggressive or high-purity media
  • Inert flow path separated from actuator by a diaphragm
  • Side mounted ports
PRODUCT OPTIONS
$224.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection TypeSide mounted ports, 1/4" - 28 UNF
  • Housing MaterialPPS
  • Power (VDC)24
  • Flow Pattern2-way, NC
  • DescriptionIsolation Valve, 2-Way, NC, Side Mounted Ports
Key Features
  • Inert flow path separated from actuator by a diaphragm
  • Side mounted ports
More About this Item
These isolation valves offer an inert, nonmetallic flow path with a solenoid actuator separated from the fluid path by a flexible diaphragm. Valves are easy to install and are coupled with low internal volume and precise flow control. Suitable for highly aggressive or high-purity media you may encounter in analytical chemistry applications. Life expectancy is 20,000,000 cycles.