Spex VapLock™ Solvent Delivery Cap w/ Viton Air Inlet Valve, Two 1/4-28 ports, 38 mm Wako; 1/EA

Representative image only.
Spex Item # EW-12018-46
Reduce chemical evaporation and exposure to hazardous vapors
  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
  • 38 mm Wako thread with two 1/4-28 ports
  • Viton® air inlet valve

PRODUCT OPTIONS

$47.75USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Cap MaterialViton® air inlet valve
  • Cap / Stopper SizeWako 38 mm
  • Cap ColorWhite
  • AutoclavableYes
  • Special Features2 x 1/4"-28 UNF(F)
  • DescriptionSolvent Delivery Cap w/ Viton Air Inlet Valve, two 1/4"-28 ports, 38 mm Wako; 1/ea

Key Features

  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
  • 38 mm Wako thread with two 1/4-28 ports
  • Viton® air inlet valve

More About this Item

Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4"-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable.

Caps with integrated air inlet valve and filter permit air to enter the bottle for vacuum displacement while solvent is pumped to the instrument. Valves are made of PTFE and 300-series SS components and feature a 50 µm PTFE filter to reduce particulate contamination. Select from FFKM, EPDM, or Viton® valve seal options.

*Does not come in contact with liquid under normal circumstances.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat