Spex VapLock™ Solvent Waste Manifold, PP, 70 mm Nalgene, 1x1/2" NPT, 3x1/4" NPT, 6x1/4-28 UNF; 1/EA
Representative image only.
Spex Item # EW-12019-30
Connect solvent waste manifold caps to waste containers for a secure closed system
  • 70 mm Nalgene connection
  • Polypropylene construction
  • 10 ports
PRODUCT OPTIONS
$236.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Cap / Stopper Size70 mm
  • Cap ColorWhite
  • Special Features1 x 1/2" NPT(F), 3 x 1/4" NPT(F), 6 x 1/4-28 UNF(F)
  • DescriptionSolvent Waste Manifold, PP, 70 mm Nalgene, 1x1/2" NPT, 3x1/4" NPT, 6x1/4-28 UNF; 1/EA
Key Features
  • 70 mm Nalgene connection
  • Polypropylene construction
  • 10 ports
More About this Item
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Bottle and carboy caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.