Cole-Parmer VapLock™ Solvent Waste Manifold, PTFE, 2" Fine, 1x1/2" NPT, 3x1/4" NPT; 1/ea
Representative image only.
Cole-Parmer Item # EW-12019-64
Connect solvent waste manifold caps to waste containers for a secure closed system
  • Choose from polypropylene or PTFE construction
PRODUCT OPTIONS
$340.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap / Stopper Size2\" Fine Thread
  • Special Features1x1/2\" NPT, 3x1/4\" NPT
  • DescriptionSolvent Waste Manifold, PTFE, 2\" Fine, 1x1/2\" NPT, 3x1/4\" NPT; 1/ea
Key Features
  • Choose from polypropylene or PTFE construction
More About this Item
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Drum caps fit on 2" bung opening (fine or coarse thread) and come with PP drum thread adapter. Caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.