Cole-Parmer VapLock™ Solvent Waste Manifold, PTFE, 63 mm Fort-Pak Container, 1x1/2" NPT, 3x1/4" NPT, 6x1/4-28 UNF; 1/EA
Representative image only.
Cole-Parmer Item # EW-12021-06
Connect solvent waste manifold caps to waste containers for a secure closed system
  • Choose from polypropylene or PTFE construction
  • 64 mm Fort-Pak container connection
  • PTFE construction
  • 10 ports
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap / Stopper Size63 mm Fort-Pak Container
  • Cap ColorWhite
  • Special Features1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
  • DescriptionSolvent Waste Manifold, PTFE, 63 mm Fort-Pak Container, 1x1/2\" NPT, 3x1/4\" NPT, 6x1/4-28 UNF; 1/EA
Key Features
  • Choose from polypropylene or PTFE construction
  • 64 mm Fort-Pak container connection
  • PTFE construction
  • 10 ports
More About this Item
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Bottle and carboy caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.