Spex VapLock™ Waste Funnel, 4" dia., Justrite Centura, SS / PTFE quick disconnect, HDPE; 1/ea
Representative image only.
Spex Item # EW-12020-65
Safely and securely capture waste and hazardous solvents
  • Reduces fumes and harmful vapor exhaust
  • Easily connect to Justrite® Centura safety containers
PRODUCT OPTIONS
$226.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSS / PTFE quick disconnect, HDPE funnel
  • Compatible WithJustrite Centura containers
  • Diameter (in)4
  • Diameter (cm)10.2
  • DescriptionWaste Funnel, 4" dia., Justrite Centura, SS / PTFE quick disconnect, HDPE; 1/ea
Key Features
  • Reduces fumes and harmful vapor exhaust
  • Easily connect to Justrite® Centura safety containers
More About this Item
Use these 4" diameter funnels to safely dispose of liquid laboratory waste. A locking hinged lid minimizes leaks and spills when the funnel is closed. Quick-disconnect fitting on models 12020-64 and -65 allows fast and easy connection to Justrite Centura safety containers. Funnels are constructed of high-density polyethylene (HDPE) and polypropylene (PP) with a Viton® gasket.