Spirax Sarco 13WS 1" 69844  Air Eliminator, Cast Iron, 1" NPT(F) x 3/8" NPT(F)
Representative image only.
Spirax Sarco Mfr # 13WS 1" 69844 – Item # EW-07600-94
  • Improve the circulation of pressurized liquids by eliminating air and other non-condensable gases
  • Mount at high points to provide immediate removal of air and gases
  • EPDM seals ensure positive shut-off
  • Cast-iron body with stainless steel internal components — can be serviced without disturbing the inlet piping connection
PRODUCT OPTIONS
$297.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port3/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1" NPT(F)
  • Max Temperature (° F)450
  • Max Temperature (° C)232
  • Wetted MaterialsCast iron
  • DescriptionAir Eliminator, Cast Iron, 1" NPT(F) x 3/8" NPT(F)
Key Features
  • Improve the circulation of pressurized liquids by eliminating air and other non-condensable gases
  • Mount at high points to provide immediate removal of air and gases
  • EPDM seals ensure positive shut-off
  • Cast-iron body with stainless steel internal components — can be serviced without disturbing the inlet piping connection
More About this Item
Typical applications: Use for both hot and cold liquid services
  • Cold water lines
  • Suction lines to pumps
  • Mixing tanks
  • Condensate return lines
  • Cool water lines on air compressors
  • Water storage tanks