Spirax Sarco B1H-125 1/2" 64150  Cast iron inverted bucket steam trap; 1/2" NPT(F), 125 psig
Representative image only.
Spirax Sarco Mfr # B1H-125 1/2" 64150 – Item # EW-07600-60
  • Keep steam space free of condensate to provide maximum system efficiency
  • All stainless steel working parts provide good resistance to corrosion and wear
  • Inverted bucket steam traps rely on the difference in density between steam and condensate to drain condensate as it forms.
  • Condensate is discharged close to steam temperature
PRODUCT OPTIONS
$233.20USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Max Pressure (PSI)125
  • Max Temperature (° F)450
  • Max Temperature (° C)232
  • Wetted MaterialsCast iron
  • DescriptionCast iron inverted bucket steam trap; 1/2" NPT(F), 125 psig
Key Features
  • Keep steam space free of condensate to provide maximum system efficiency
  • All stainless steel working parts provide good resistance to corrosion and wear
  • Inverted bucket steam traps rely on the difference in density between steam and condensate to drain condensate as it forms.
  • Condensate is discharged close to steam temperature
More About this Item
Typical applications:
  • Steam main drip stations
  • Laundry equipment
  • Industrial dryers
  • Storage tanks