Spirax Sarco FT-15 3/4" 50926  Cast Iron Float/Thermostatic Steam Trap, Parallel Config.; 3/4"NPT(F), 15 psig
Representative image only.
Spirax Sarco Mfr # FT-15 3/4" 50926 – Item # EW-07600-30
$362.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialCast iron
  • Connection TypeNPT(F)
  • Thread size NPT(F) (port 1) (in)3/4
  • Max Temperature (° F)450°F of superheat
  • Max Temperature (° C)232°C of superheat
  • Max Operating Pressure (PSI)15
  • DescriptionCast Iron Float/Thermostatic Steam Trap, Parallel Config.; 3/4"NPT(F), 15 psig
More About this Item
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections.
Typical applications:
  • All process equipment, particularly when controlled by modulating temperature control valves
  • Unit heaters
  • Air heat coils
  • Heat exchangers
  • Steam main drip stations