Spirax Sarco TD52 3/8" 54529C Thermo-Dynamic Steam Trap, 3/8"NPT(F), 600 psi
Representative image only.
Spirax Sarco Mfr # TD52 3/8" 54529C – Item # EW-07600-02
$402.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Connection TypeNPT(F)
  • Thread size NPT(F) (port 1) (in)3/8
  • Thread size NPT(F) (port 2) (in)3/8
  • Max Temperature (° F)800
  • Max Temperature (° C)427
  • Max Operating Pressure (PSI)600
  • DescriptionThermo-Dynamic Steam Trap, 3/8"NPT(F), 600 psi
More About this Item
Typical applications:
  • Steam main drain and tracer lines
  • Process equipment
  • Laundry and kitchen equipment
  • Superheated steam applications
  • Outdoor installations subject to freezing