Spirax Sarco Thermo-Dynamic steam trap with blowdown valve, 1/2" NPT(F)
Representative image only.
Spirax Sarco Mfr # TD42L 1/2" 0680762 – Item # EW-07600-10
Keep steam space free of condensate to provide maximum system efficiency
  • Constructed of durable stainless steel (SS)
$765.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Connection TypeNPT(F)
  • Thread size NPT(F) (port 1) (in)1/2
  • Thread size NPT(F) (port 2) (in)1/2
  • Max Temperature (° F)750
  • Max Temperature (° C)400
  • Max Operating Pressure (PSI)600
  • DescriptionThermo-Dynamic steam trap with blowdown valve, 1/2" NPT(F)
Key Features
  • Constructed of durable stainless steel (SS)
More About this Item
Includes strainer for added protection from dirt and scale.
Typical applications:
  • Steam main drainage
  • Tracer lines
  • Process equipment
  • Outdoor installations subject to freezing