Spring tubing clamp for up to 1/2"OD tubing, 3/bag
Representative image only.
Item # EW-06834-20
An ideal option to secure tubing
  • Constructed of durable acetal copolymer and stainless steel
PRODUCT OPTIONS
$46.50USD / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialAcetal copolymer and stainless steel
  • Clamp TypeTubing
  • Min Grip Range (in)0.313
  • Max Grip Range (in)0.5
  • DescriptionSpring tubing clamp for up to 1/2"OD tubing, 3/bag
Key Features
  • Constructed of durable acetal copolymer and stainless steel
More About this Item
Acetal copolymer and stainless steel clamps provide on/off flow control for flexible tubing. Chemically resistant. Bel-Art. 12 bg/cs.