Standard methods agar, prepared; 200 mL bottle, 12/pk
Representative image only.
Item # EW-14204-88
Save time and money with ready-made bottles
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • Plate count agar for the cultivation and enumeration of microorganisms in dairy products, food, and water
PRODUCT OPTIONS
$158.00USD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Standard Methods
  • Capacity (mL)200
  • Capacity (L)0.2
  • DescriptionStandard methods agar, prepared; 200 mL bottle, 12/pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • Plate count agar for the cultivation and enumeration of microorganisms in dairy products, food, and water
More About this Item
Bottles are tested and standardized to ensure consistent and reliable performance. Each bottle includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Bottle and pack sizes vary.